Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Anisil 98.0+%, TCI America™
SDP

Catalog No. A102825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A102825G 25 g
A10285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A102825G Supplier TCI America Supplier No. A102825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1226-42-2
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.284
MDL Number MFCD00008405
InChI Key YNANGXWUZWWFKX-UHFFFAOYSA-N
Synonym 4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione
PubChem CID 71043
IUPAC Name 1,2-bis(4-methoxyphenyl)ethane-1,2-dione
SMILES COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC

Specifications

Melting Point 133°C
Quantity 25 g
Formula Weight 270.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Anisil
EINECSNumber (4)-0277
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.