Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Benzoquinone, ≥99.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018250 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8250 100 g
11-101-9219 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8250 Supplier MilliporeSigma Supelco Supplier No. 12309100G
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00001591
Synonym Quinone

Specifications

Melting Point 112°C to 114°C; 113°C to 115°C
CAS 106-51-4
Ignition Point 815°F
Linear Formula C6H4(=O)2
Quantity 100 g
Vapor Density 3.73 (vs. air)
Formula Weight 108.09
Vapor Pressure 0.1 mmHg (25°C)
Percent Purity ≥99.5% (HPLC)
RTECSNumber DK2625000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.