Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P-Cadherin Monoclonal Antibody (CSTEM29), APC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 17223742
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17223742 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17223742 Supplier Invitrogen™ Supplier No. 17223742
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

This monoclonal antibody CSTEM29 reacts with human P-Cadherin, also known as Cadherin-3. Applications Reported: This CSTEM29 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CSTEM29 antibody has been pre-diluted and tested by flow cytometric analysis of normal human induced pluripotent stem cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

P-cadherin, also known as Cadherin-3, is a Type 1 cadherin protein that belongs to the cadherin superfamily. Type 1 cadherins are single-pass transmembrane proteins that have 5 extracellular cadherin repeats and an intracellular domain that binds p120-catenin and beta-catenin. Cadherins are calcium-dependent cell-cell adhesion glycoproteins responsible for a range of processes including development, wound healing, cell-cell signaling, cell growth and differentiation. P-cadherin is expressed in human fetal structures, and in adult tissues such as the basal layer of the epidermis, breast, prostate, ovary, cervix, hair follicle, and corneal endothelium. P-cadherin expression has also been reported on embryonic stem cells, and stem cells of the normal mammary gland, and hair follicle. Overexpression of P-cadherin has been associated with poor prognosis in breast, prostate, ovary, colon, and stomach carcinomas. Mutations in CDH3 are associated with congential hypotrichosis with juvenile macular dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P-Cadherin
Applications Flow Cytometry
Classification Monoclonal
Clone CSTEM29
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Cdh3
Gene Accession No. P22223
Gene Alias AI385538; CADH3; cadherin 15, M-cadherin (myotubule); cadherin 3; cadherin 3, type 1, P-cadherin (placental); cadherin-3; Cadp; calcium-dependent adhesion protein, placental; CDH3; CDHP; HJMD; PCAD; P-cadherin; Placental cadherin; Placental-cadherin; RPE-specific cadherin
Gene Symbols Cdh3
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1001
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.