Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Cresol (Laboratory), Fisher Chemical

Catalog No. O20434 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O2043-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O2043-4

missing translation for 'validMainframeMsg'

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1319-77-3
Molecular Formula C7H8O
MDL Number MFCD00002376
Synonym 4-Methyl Phenol, Cresylic Acid

Specifications

Melting Point 12°C
Color Yellow to Orange
Density 1.0400g/cm³
Packaging Glass Bottle
Linear Formula CH3C6H4OH
Quantity 4 L
Vapor Density 3.7g/mol
Formula Weight 108.14
Grade Laboratory
Physical Form Liquid
Chemical Name or Material p-Cresol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.