Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Cresol, Puriss p.a., ≥99.0% (GC),Solstice

Catalog No. 60048531 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-531 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-531 Supplier Solstice Supplier No. 6103025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 106-44-5
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002376
InChI Key IWDCLRJOBJJRNH-UHFFFAOYSA-N
Synonym p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
PubChem CID 2879
ChEBI CHEBI:17847
IUPAC Name 4-methylphenol
SMILES CC1=CC=C(C=C1)O

Specifications

Melting Point 32°C to 35°C
Density 1.034g/mL at 25°C
Boiling Point 202°C
Flash Point 85°C (185°F)
Ignition Point 1038°F
Packaging Glass bottle
Linear Formula CH3C6H4OH
Quantity 25 g
UN Number UN3455
Beilstein 1305151
Vapor Density 3.72 (vs air)
Formula Weight 108.14g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material p-Cresol
Show More Show Less
Hazard Statement P280-P301 + P310-P305 + P351 + P338-P310
DOTInformation 8(6.1)/PGII
RTECSNumber 203-398-6
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.