Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ p-Dichlorobenzene
SCI_ED

Catalog No. S25299B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
3 kg
Packaging:
Lab Grade
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25299B 3 kg Lab Grade
S25299 500 g Lab Grade
S25299C 2.5 kg Lab Grade
S25299A 1 kg Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. S25299B
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 106-46-7
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
InChI Key OCJBOOLMMGQPQU-UHFFFAOYSA-N
Synonym p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
PubChem CID 4685
ChEBI CHEBI:28618
IUPAC Name 1,4-dichlorobenzene
SMILES C1=CC(=CC=C1Cl)Cl

Specifications

Color White
Density 1.458g/mL at 20°C
Packaging Lab Grade
Linear Formula C6H4Cl2
Quantity 3 kg
Formula Weight 147
Grade Laboratory
Chemical Name or Material p-Dichlorobenzene
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 9, III, : Miscellaneous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.