Learn More
p-Dimethylaminobenzalrhodanine, 0.02% (w/v) in Acetone, Indicator for Argentimetric Titrations, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| CAS Min % | 98.0, 0.02 |
| CAS Max % | 100.0, 0.03 |
| Concentration | 0.02% (w/v) |
| Grade | Indicator |
| Packaging | Amber Glass Bottle |
| Solvent or Matrix | Acetone |
| Color | Orange |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Chemical Name or Material | p-Dimethylaminobenzalrhodanine, Indicator For Argentimetric Titrations |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.