Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Dimethylaminobenzoic Acid, MP Biomedicals

Catalog No. ICN205110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205110 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205110 Supplier MP Biomedicals, Inc Supplier No. 0520511025

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 619-84-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
InChI Key YDIYEOMDOWUDTJ-UHFFFAOYSA-N
Synonym 4-dimethylamino benzoic acid, benzoic acid, 4-dimethylamino, benzoic acid, p-dimethylamino, p-dimethylaminobenzoic acid, n,n-dimethyl-p-aminobenzoic acid, n,n-dimethyl-4-aminobenzoic acid, p-n,n-dimethylamino benzoic acid, p-dimethylamino benzoic acid, unii-d1ma908ev0, 4-n,n-dimethylamino benzoic acid
PubChem CID 12092
IUPAC Name 4-(dimethylamino)benzoic acid
SMILES CN(C)C1=CC=C(C=C1)C(=O)O

Specifications

Quantity 100 g
Formula Weight 165.19
Chemical Name or Material p-Dimethylaminobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.