Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Fluorobenzenesulfonic Acid, Na Salt,MP Biomedicals

Catalog No. ICN204870 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204870 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204870 Supplier MP Biomedicals, Inc Supplier No. 0520487001

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 368-88-7
Molecular Formula C6H5FO3S
Molecular Weight (g/mol) 176.16
MDL Number MFCD06658253
InChI Key WVSYONICNIDYBE-UHFFFAOYSA-N
Synonym 4-fluorobenzenesulphonic acid, 4-fluoro-benzenesulfonic acid, benzenesulfonic acid, 4-fluoro, benzenesulfonic acid,4-fluoro, p-fluorobenzenesulfonic acid, zlchem 971, 4-fluorobenzenesulphonicacid, 4-fluoranylbenzenesulfonic acid, 4-fluoro-benzene-sulfonic acid, p-fluorobenzensulfonic acid
PubChem CID 46886
IUPAC Name 4-fluorobenzene-1-sulfonic acid
SMILES OS(=O)(=O)C1=CC=C(F)C=C1

Specifications

Quantity 10 g
Chemical Name or Material p-Fluorobenzenesulfonic Acid, Na Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.