Learn More
Medchemexpress LLC P-MPPI hydrochloride | 220643-77-6 | 99.8% | 10 MG

Supplier: Medchemexpress LLC HY12073810MG
p-MPPI hydrochloride is a selective 5-HT1A receptor antagonist used in preclinical research to study serotonin receptor pharmacology and behavioral effects. It is supplied as a high-purity solid for analytical and in vivo studies.
- Selective 5-HT1A receptor antagonist for pharmacology studies.
- High purity suitable for analytical and preclinical applications.
- Solid, white to off-white appearance for easy handling.
- CAS number 220643-77-6 for unambiguous identification.
- Molecular weight 578.87 g/mol; formula C25H28ClIN4O2.
- Store sealed, protected from moisture and light.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.