Learn More
Sigma Organic Chemistry P,P-Dichlorophenylphosphine | 100G | 644-97-3 | MFCD00000528
Supplier: Sigma Organic Chemistry D71984100G

P,P-Dichlorophenylphosphine, 100G
About This Item:
Synonym(s): Phenyldichlorophosphine; Phenylphosphine dichloride; PhPCl2; Phenylphosphonous dichloride
Linear Formula: C6H5PCl2
Molecular Weight: 178.98
CAS Number: 644-97-3
MDL Number: MFCD00000528
UNSPSC Code: 12352002
Purity: 0.97
Boiling Point: 222 C/759 mmHg (lit.)
Melting Point: -51 C (lit.)
Density: 1.319 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.597 (lit.)
Storage: room temp
EINECS Number: 211-425-8
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.