Learn More
ABclonal Technology Phospho-GCN2-T899 Rabbit mAb
SDP
Supplier:  ABclonal Technology AP1356100UL
Encompass_Preferred
This gene encodes a member of a family of kinases that phosphorylate the alpha subunit of eukaryotic translation initiation factor-2 (EIF2) resulting in the downregulaton of protein synthesis The encoded protein responds to amino acid deprivation by binding uncharged transfer RNAs It may also be activated by glucose deprivation and viral infection Mutations in this gene have been found in individuals suffering from autosomal recessive pulmonary venoocclusive-disease-2
Catalog No. 50-000-14084
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.