Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Quinone-4-chloroimide 95.0+%, TCI America™
SDP

Catalog No. Q00191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00191G 1 g
Q00195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00191G Supplier TCI America Supplier No. Q00191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 637-61-6
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.554
MDL Number MFCD00058961
InChI Key ITUYMTWJWYTELW-UHFFFAOYSA-N
Synonym N-Chloro-p-benzoquinone Monoimine, N-Chloro-p-quinone Monoimine
PubChem CID 12505
IUPAC Name 4-chloroiminocyclohexa-2,5-dien-1-one
SMILES C1=CC(=O)C=CC1=NCl

Specifications

Melting Point 85°C
Quantity 1 g
UN Number 2811
Formula Weight 141.55
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material p-Quinone-4-chloroimide
RTECSNumber GU5434000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.