Learn More
Invitrogen™ P-Selectin Monoclonal Antibody (CRC81), PE

Description
P-selectin (Selectin P, GMP-140, LECAM3, CD62 antigen-like family member) is a 140 kDa type 1 transmembrane glycoprotein, and is one of the most commonly studied proteins that regulate cell rolling. P-selectin is stored in the Weibel-Palade bodies of endothelial cells, as well as in a-granules of platelets. From there, it can be rapidly brought to the cell surface after exposure to thrombin, histamine, complement 5a, Ca21 ionophores, or adenosine diphosphate.
Specifications
Specifications
| Antigen | P-Selectin |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CRC81 |
| Concentration | Conc. Not Determined |
| Conjugate | PE |
| Formulation | PBS with 4mg/mL BSA and 0.1% sodium azide |
| Gene | SELP |
| Gene Accession No. | P16109 |
| Gene Alias | CD62; CD62 antigen-like family member P; CD62P; CD62P antigen; CD62P; CD62 homolog; FLJ45155; GMP140; GMP-140; GMRP; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; Grmp; LECAM3; leukocyte-endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; PSEL; PSELECT; P-selectin; P-selectin precursor; RP1-86F14.2; selectin P; selectin P (granule membrane protein 140kDa, antigen CD62); selectin, platelet; Selp |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.