Learn More
Medchemexpress LLC P-Selectin Mouse H 10ug
SDP
Supplier:  Medchemexpress LLC HYP7338310UG
Encompass_Preferred
P-selectin is a Ca(2 )-dependent receptor on myeloid cells that specifically binds to carbohydrates on neutrophils and monocytes It mediates interactions between activated endothelial cells or platelets and leukocytes by recognizing sialic acid-Lewis X P-Selectin Protein Mouse (HEK293 Fc) is the recombinant mouse-derived P-Selectin protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-79420