Learn More
ABclonal Technology p-Smad2/3-S465/S467/S423/S425
SDP
Supplier:  ABclonal Technology AP134320UL
Encompass_Preferred
The protein encoded by this gene belongs to the SMAD family which includes proteins similar to Drosophila Mad and C elegans Sma SMAD proteins are signal transducers and transcriptional modulators involved in multiple signaling pathways This protein mediates the transforming growth factor (TGF)-beta signal regulating cell processes like proliferation apoptosis and differentiation It is recruited to TGF-beta receptors via its interaction with the SMAD anchor for receptor activation (SARA) protein Phosphorylation by TGF-beta receptors causes dissociation from SARA and association with SMAD4 which is essential for nuclear translocation In the nucleus it binds to target promoters and forms a transcription repressor complex The protein can also be phosphorylated by activin type 1 receptor kinase mediating activin signaling Alternative splicing results in multiple transcript variants
Catalog No. 50-000-14191
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.