Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolualdehyde, 99+%

Catalog No. AC421140025
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421140025 2.5 kg Glass bottle
AC421140050 5 g Glass bottle
AC421141000 100 g Glass bottle
AC421145000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC421140025 Supplier Thermo Scientific Chemicals Supplier No. 421140025

CAS: 104-87-0 | C8H8O | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-87-0
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00006954
InChI Key FXLOVSHXALFLKQ-UHFFFAOYSA-N
Synonym p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
PubChem CID 7725
ChEBI CHEBI:28617
IUPAC Name 4-methylbenzaldehyde
SMILES CC1=CC=C(C=C1)C=O

Specifications

Melting Point -6.0°C
Color Colorless to Yellow
Density 1.0150g/mL
Boiling Point 204.0°C to 205.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5420 to 1.5480
Linear Formula CH3C6H4CHO
Quantity 2.5 kg
Beilstein 07,297
Specific Gravity 1.015
Solubility Information Solubility in water: 0.25g/l (25°C). Other solubilities: soluble in alcohol and ether
Formula Weight 120.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material p-Tolualdehyde, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 203-246-9
RTECSNumber CU7034500
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.