Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonyl Chloride p.a., Thermo Scientific™

Catalog No. AC220541000
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC220541000 100 g
AC220540100 10 g
AC220545000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC220541000 Supplier Thermo Scientific Chemicals Supplier No. 220541000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-59-9
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.64
MDL Number MFCD00007450
InChI Key YYROPELSRYBVMQ-UHFFFAOYSA-N
Synonym tosyl chloride, p-toluenesulfonyl chloride, p-tosyl chloride, 4-toluenesulfonyl chloride, p-toluenesulfochloride, 4-methylbenzene-1-sulfonyl chloride, p-tolylsulfonyl chloride, p-toluenesulphonyl chloride, tosylchloride, p-toluenesulfonic acid chloride
PubChem CID 7397
IUPAC Name 4-methylbenzene-1-sulfonyl chloride
SMILES CC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Color White
Infrared Spectrum Authentic
Linear Formula CH3C6H4SO2Cl
Quantity 100 g
Beilstein 11, 103
Fieser 01,1179; 03,292; 04,510; 05,676; 06,598; 08,489; 11,536; 13,286
Merck Index 15, 9693
Formula Weight 190.64
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material p-Toluenesulfonyl chloride, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement MOISTURE SENSITIVE
EINECSNumber 202-684-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.