Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonyl isocyanate, 96%

Catalog No. p-3737556
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC183070250 25 mL Glass bottle
AC183071000 100 mL Glass bottle
AC183075000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC183070250 Supplier Thermo Scientific Chemicals Supplier No. 183070250
Only null left
Add to Cart

CAS: 4083-64-1 | C8H7NO3S | 197.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4083-64-1
Molecular Formula C8H7NO3S
Molecular Weight (g/mol) 197.21
MDL Number MFCD00002030
InChI Key VLJQDHDVZJXNQL-UHFFFAOYSA-N
Synonym tosyl isocyanate, p-toluenesulfonyl isocyanate, 4-methylbenzenesulfonyl isocyanate, p-toluenesulphonyl isocyanate, benzenesulfonyl isocyanate, 4-methyl, tosylisocyanate, p-tolylsulfonyl isocyanate, unii-h9004fjx6v, p-toluenesulfonic acid, anhydride with isocyanic acid, p-toluene sulfonyl isocyanate
PubChem CID 77703
IUPAC Name 4-methyl-N-(oxomethylidene)benzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N=C=O

Specifications

Melting Point -2°C
Color Colorless to Yellow
Density 1.2900g/mL
Boiling Point 144°C (10.0 mmHg)
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5330 to 1.535
Linear Formula CH3C6H4SO2NCO
Quantity 25 mL
Fieser 07,377
Specific Gravity 1.29
Solubility Information Solubility in water: reacts
Formula Weight 197.21
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material p-Toluenesulfonyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-810-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.