Learn More
Medchemexpress LLC p-Toluic acid-d7 | 1219798-76-1 | MFCD06658857 | 99.7% | 10 MG
Supplier: Medchemexpress LLC HY76547S110MG
p-Toluic acid-d7 is the deuterium-labeled form of p-toluic acid (4-methylbenzoic acid), provided as a stable-isotope reference standard for analytical chemistry, metabolite tracing, and mass spectrometry applications. CAS 1219798-76-1; molecular weight 143.19 g/mol. Certificate of analysis reports high chemical purity and isotopic enrichment.
- High isotopic enrichment for accurate mass spectrometry.
- High chemical purity (~99.7% as reported by COA).
- Small 10 mg size suitable for method development and standards.
- Supplied with certificate of analysis and safety data sheet for traceability.
- Stable solid form, convenient for storage and handling.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.