Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolyl Isobutyrate 97.0+%, TCI America™
SDP

Catalog No. I039625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I039625ML 25 mL
I03965ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I039625ML Supplier TCI America Supplier No. I039625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 103-93-5
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00026440
InChI Key UPPSFGGDKACIKP-UHFFFAOYSA-N
Synonym Isobutyric Acid p-Tolyl Ester
PubChem CID 7685
IUPAC Name 4-methylphenyl 2-methylpropanoate
SMILES CC(C)C(=O)OC1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 237°C
Quantity 25 mL
Formula Weight 178.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Tolyl Isobutyrate
RTECSNumber NQ5460000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.