Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. I039325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I039325G 25 g
I03935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I039325G Supplier TCI America Supplier No. I039325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 622-59-3
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00004813
InChI Key ABQKHKWXTUVKGF-UHFFFAOYSA-N
Synonym p-tolyl isothiocyanate, 4-methylphenyl isothiocyanate, 4-methylphenylisothiocyanate, p-tolylisothiocyanate, 4-tolylisothiocyanate, benzene, 1-isothiocyanato-4-methyl, 4-tolyl isothiocyanate, isothiocyanic acid, p-tolyl ester, 1-isothiocyanato-4-methyl-benzene, isothiocyanic acid, 4-tolyl ester
PubChem CID 12149
IUPAC Name 1-isothiocyanato-4-methylbenzene
SMILES CC1=CC=C(C=C1)N=C=S

Specifications

Color White-Yellow
Boiling Point 237°C
Quantity 25 g
UN Number 2811
Formula Weight 149.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material p-Tolyl Isothiocyanate
RTECSNumber NX9500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.