Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylene-2-sulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D068325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D068325G 25 g
D06835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D068325G Supplier TCI America Supplier No. D068325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19040-62-1
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD00024875
InChI Key FZVZUIBYLZZOEW-UHFFFAOYSA-N
Synonym p-xylene-2-sulfonyl chloride, benzenesulfonyl chloride, 2,5-dimethyl, 2,5-dimethylbenzene-1-sulfonyl chloride, 2,5-dimethylphenylsulfonyl chloride, 2,5-dimethyl-benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-dimethyl, 2,5-dimethylphenyl chlorosulfone, acmc-209etq, xylenesulfonylchloride, ksc178m0f
PubChem CID 87910
IUPAC Name 2,5-dimethylbenzenesulfonyl chloride
SMILES CC1=CC(=C(C=C1)C)S(=O)(=O)Cl

Specifications

Color White-Yellow
Quantity 25 g
Physical Form Crystal at 20°C
UN Number 3261
Boiling Point 150°C
Formula Weight 204.67
Percent Purity ≥98.0% (T)
Chemical Name or Material p-Xylene-2-sulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.