Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylene, ≥99% (GC), Solstice

Catalog No. 60048570 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-570 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-570 Supplier Solstice Supplier No. 956822.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 106-42-3 | C8H10 | 106.17 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-42-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008556
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
PubChem CID 7809
ChEBI CHEBI:27417
IUPAC Name 1,4-xylene
SMILES CC1=CC=C(C)C=C1

Specifications

Melting Point 12-13°C
Density 0.861g/mL at 20°C
Quantity 2.5 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Ignition Point 984°F
Packaging Glass Bottle
Refractive Index 1.497
Linear Formula C6H4(CH3)2
Vapor Pressure 9mmHg (20°C)
UN Number UN1307
Beilstein 1901563
Boiling Point 138°C to 139°C
Formula Weight 106.17g/mol
Percent Purity ≥99.0% (GC)
Grade Not Available
Chemical Name or Material p-Xylene
Show More Show Less
Hazard Statement P280
DOTInformation class 3 - PG 3 - Xylenes
RTECSNumber 203-396-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.