Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylylene Dicyanide 98.0+%, TCI America™
SDP

Catalog No. X006125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X006125G 25 g
X00615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X006125G Supplier TCI America Supplier No. X006125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 622-75-3
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00001923
InChI Key FUQCKESKNZBNOG-UHFFFAOYSA-N
Synonym 1,4-phenylenediacetonitrile, 1,4-benzenediacetonitrile, p-xylylene dicyanide, p-phenylenediacetonitrile, p-benzenediacetonitrile, 2,2'-1,4-phenylene diacetonitrile, p-bis cyanomethyl benzene, 1,4-bis cyanomethyl benzene, p-cyanomethyl benzyl cyanide, 2-4-cyanomethyl phenyl acetonitrile
PubChem CID 69327
IUPAC Name 2-[4-(cyanomethyl)phenyl]acetonitrile
SMILES C1=CC(=CC=C1CC#N)CC#N

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 156.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Xylylene Dicyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.