Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hypochlorite Solution (4-6%/Laboratory), Fisher Chemical™
GSA_VA

Catalog No. SS290-1 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SS290-1 1 L Poly Bottle
SS290-4 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SS290-1
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9 , 7732-18-5
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Color Yellow
Melting Point 0°C
Density 1.1g/cm³
Formula Weight 74.44
Percent Purity 4 to 6%
Grade Laboratory
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 14mmHg
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Sodium Hypochlorite Solution, 4-6%
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.