Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea (Certified ACS), Fisher Chemical™

Catalog No. U15-500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
3 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
U15-500 500 g Poly Bottle
U15-3 3 kg Poly Bottle
U15-50 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. U15-500
  • $188.00
    $155.65 / Each

    Save $32.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $896.00
    $757.65 / Case of 6 EA

    Save $138.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Decomposition Information Decomposes information: >132°C
Melting Point 131°C
Color White
Density 1.335g/cm³
pH 7.5 to 9.5
Ignition Residue 0.01% max.
Assay Percent Range 99.0 to 100.5 %
Packaging Poly Bottle
Linear Formula NH2CONH2
Quantity 500 g
Solubility Information 0.01% max. Insoluble Matter
Vapor Pressure 1.25mmHg at 25°C
Formula Weight 60.06
Percent Purity 99.0 to 100.5%
Grade Certified ACS
Physical Form Solid
Chemical Name or Material Urea
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation.
NFPA
Health:1 Flammability:0 Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.