Learn More
Velp™ P0 Glass Crucibles for FIWE3 Fiber Analyzer
SDP
Catalog No. 11392028
Change view
Click to view available options
Porosity:
Class 3
Catalog No. Porosity
11-392-028 Class 3
1 options
Catalog No. 11-392-028 Supplier Velp™ Supplier No. A00000138
Add to Cart
Add to Cart
P0 glass crucibles for use with the FIWE3 Fiber Analyzer.
  • Glass crucibles, P0
  • For fiber analyzer
Specifications
Material Glass
Quantity 6 Crucibles
Capacity (Metric) 160 to 250 microns
Autoclavable Not Autoclavable
Product Type Glass Crucible
Porosity Class 3
Shape Round