Learn More
Apexbio Technology LLC P005091, 50mg. Cas: 882257-11-6 MFCD: MFCD00123202. Ubiquitin-specific protease 7 (USP7) inhibitor
SDP
Supplier:  Apexbio Technology LLC A302350
Encompass_Preferred
P005091 is a selective, potent inhibitor of USP7 deubiquitylating activity, which is demonstrated to downregulate claspin and phosphorylation of the DNA checkpoint kinase Chk1, and to synergize with genotoxic agents Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4785
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.