Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Pentobarbital Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco P0101ML

Catalog No. 01-001-826


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Pentobarbital is a short-acting barbiturate and hypnotic approved for treatment of seizures and preoperative sedation. Also known as pentobarbitone, the sodium salt of pentobarbital is used as an anesthetic in veterinary medicine. This certified solution standard is suitable for GC/MS and LC/MS testing applications in forensic analysis, clinical toxicology or urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1, 76-74-4
UN 1230 - class 3 - PG 2 - Methanol, solution
Ampule
9.7°C (49.5°F) - closed cup
MFCD00057561
1.0 mg/mL in methanol
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

CERILLIANT is a registered trademark of Cerilliant Corporation