Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ P100 Low Profile Cartridge
SDP

Catalog No. 19222501 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Quantity:
1 Pair
100/Cs.
6/box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-222-501 100/Cs.
19-222-502 1 Pair
19-222-500 6/box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-222-501 Supplier Dentec Safety™ Supplier No. 158TLP0B100
Add to Cart
Add to Cart

Dentec Safety™ P100 Low Profile Cartridge is uniquely designed with a space plenum between the filtering material and the inhalation port

  • Inhaled air is evenly distributed across the filter, lowering breathing resistance and extending cartridge life.
  • Ultra thin design fits under any grinding shield or welding helmet.
  • Will not obstruct field of vision
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
  • Offset connector improves visibility and allows face shields or welding helmets to be worn without interference

Specifications

Color Purple
Material Plastic
Product Type Respirator Accessories
For Use With (Equipment) Masks
Includes Cartridges and filters
Quantity 100/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.