Learn More
Newcomer Supply p16 Control Slides, 10/set
SDP
Supplier:  Newcomer Supply 3625A
Encompass
Newcomer Supply p16 Control Slides are for the positive immunohistochemical staining of p16, a tumor suppressor protein that plays an important role in cell cycle regulation and acts as a tumor suppressor implicated in the prevention of cancers. p16 can serve as a surrogate marker for high risk HPV in cases of cervical, head and neck and a variety of HPV related carcinomas. p16 Control Slides contain sections of positive staining breast and negative staining adipose.
Catalog No. NC2101948
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.