Learn More
Enzo Life Sciences p16INK4a monoclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS3770100
Encompass_Preferred
Isotype: IgG. Immunogen: Recombinant p16. Species reactivity: Human. Applications: IHC, WB. Purity: Affinity-purified. Formulation: Liquid. In Tris buffer, pH 7.5, containing 1% BSA and <0.1% sodium azide.
Catalog No. 50-201-2904
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.