Learn More
Description
A suggested positive control is mouse colon tissue lysate. PA5-20379 can be used with blocking peptide PEP-0496. Antibodies should not be exposed to prolonged high temperatures.
Specifications
Specifications
| Antigen | p16INK4a |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CDKN2A |
| Gene Accession No. | P42771, Q64364, Q8QZZ9 |
| Gene Alias | Alternative reading frame; Arf; ARF-INK4a; CDK4 inhibitor p16 INK4; CDK4 inhibitor p16-INK4; CDK4I; CDKN2; Cdkn2a; cdkn2a {ECO:0000312; cell cycle inhibitor; cell cycle negative regulator beta; cell cycle regulator; CMM2; Cyclin dependent kinase 4 inhibitor A; cyclin dependent kinase inhibitor 2A; Cyclin dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2a p16Ink4a; cyclin-dependent kinase inhibitor 2a p19Arf; cyclin-dependent kinase inhibitor 2A, isoform 1; cyclin-dependent kinase inhibitor 2A, isoform 2; cyclin-dependent kinase inhibitor 2A, isoform 3; cyclin-dependent kinase inhibitor 2A, isoforms 1/2; cyclin-dependent kinase inhibitor protein; EMBL:AAL76336.1, ECO:0000312; EMBL:AAL76338.1, ECO:0000312; HGNC:1787; inhibits CDK4; INK4; INK4A; Ink4a/Arf; INK4a-ARF; isoforms 1/2/3; Melanoma p16 inhibits CDK4; mitochondrial smARF; MLM; MTS1; MTS-1; Multiple tumor suppressor 1; P14; P14 P16; P14ARF; p16; p16(INK4a); p16Cdkn2a; P16INK4; p16-INK4; p16INK4a; p16-INK4a; P19; P19 TP16; p19>ARF>; P19ARF; Pctr1; RGD:2323}; TP16; Tumor suppressor ARF |
| Gene Symbols | CDKN2A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
