Learn More
p190RhoGEF Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
Specifications
Specifications
| Antigen | p190RhoGEF |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | ARHGEF28 |
| Gene Accession No. | Q8N1W1 |
| Gene Alias | 190 kDa guanine nucleotide exchange factor, p190RhoGEF, p190-RhoGEF, RGNEF, Rho guanine nucleotide exchange factor, Rho guanine nucleotide exchange factor (GEF) 28, Rho interacting protein 2, rho-guanine nucleotide exchange factor, RIP2 |
| Gene Symbols | ARHGEF28 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.