Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2RX5 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2A2A6, Proteintech
SDP

Catalog No. 501736857 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6857 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6857 Supplier Proteintech Group Inc Supplier No. 664301IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mmgt2 mediates Mg(2+) transport and may provide regulated pathways for Mg(2+) transport in the Golgi and post-Golgi organelles of epithelium-derived cells. Magnesium is an essential metal, but few selective transporters have been identified. The two separate gene products, Mmgt1 and Mmgt2, comprise the family that we have termed membrane Mg(2+) transporters (MMgTs), and the proteins reside in the membrane and mediate Mg(2+) transport. The transport of Mg(2+) is saturable and studies have identified that Mmgt2 fusion proteins reside in the Golgi complex and post-Golgi vesicles, including the early endosomes in COS-7 cells transfected with the respective tagged constructs. Among related pathways for Mmgt2 are transport and binding events, in particular, transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds.

Specifications

Antigen P2RX5
Applications Western Blot
Classification Monoclonal
Clone 2A2A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene P2RX5
Gene Accession No. P51578, Q93086
Gene Alias ATP receptor, P2RX5, P2X purinoceptor 5, P2X5, P2X5R, Purinergic receptor
Gene Symbols P2RX5
Host Species Mouse
Immunogen P2RX5 Fusion Protein Ag5297
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100519563, 113995, 5026, 94045
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.