Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X3-Pure Rabbit anti-Human, Non-human Primate, Rat, Polyclonal, Neuromics™

Catalog No. 502100052
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-0052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-210-0052 Supplier Neuromics, Inc Supplier No. RA14139100UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P2X3-Pure Polyclonal antibody specifically detects P2X3-Pure in Human, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot

Specifications

Antigen P2X3-Pure
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene P2rx3
Gene Accession No. P49654, P56373
Gene Alias 4930513E20Rik; ATP receptor; MGC129956; P2RX3; P2X purinoceptor 3; p2X receptor 3; P2X receptor, subunit 3; P2X3; p2X3 purinoceptor; p2xr3; Purinergic receptor; purinergic receptor P2X 3; purinergic receptor P2X, ligand gated ion channel, 3; purinergic receptor P2X, ligand-gated ion channel, 3; purinergic receptor P2X3; purinoceptor P2X3
Host Species Rabbit
Immunogen VEKQSTDSGAYSIGH
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5024, 81739
Target Species Human, Non-human Primate, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.