Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ P2X4 Antibody Blocking Peptide
SDP

Catalog No. NBP259721 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259721 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259721 Supplier Novus Biologicals™ Supplier No. NBP259721
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide from P2X4. Source: Synthetic Amino Acid Sequence: (Accession #: NP_035156.2) C-ETDSVVSSVTTKAK The P2X4 Blocking Peptide is derived from Synthetic. The P2X4 Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Concentration LYOPH
For Use With (Application) Antibody Competition
Formulation Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt.
Gene ID (Entrez) 5025
Name P2RX4 Peptide
Quantity 0.1 mg
Storage Requirements Store at -20 C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias ATP receptor, ATP-gated cation channel protein, P2RX4
Gene Symbol P2RX4
Product Type Peptide
Reconstitution Reconstitute in 200 ul distilled water to obtain a 0.5 mg/mL peptide solution.
Research Category Signal Transduction
Purity or Quality Grade >50%, by HPLC
Protein P2X4
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.