Learn More
Specifications
Specifications
| Gene ID (Entrez) | 5027 |
| Species | Human |
| Content And Storage | Store at -20°C. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-37775 |
| Gene Alias | ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A |
| Gene Symbol | P2RX7 |
| Quantity | 0.1 mg |
| Research Category | Immunology, Signal Transduction |
| Storage Buffer | Lyophilised peptide, formulation 5 pmol/uL in 5% (v/v) acetonitrile/H2O |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
