Learn More
Invitrogen™ P2Y6 Recombinant Rabbit Monoclonal Antibody (JG37-77)

Description
Positive Control: SK-Br-3, A549, human liver tissue, human colon tissue, human kidney tissue.
Specifications
Specifications
| Antigen | P2Y6 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | JG37-77 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | P2ry6 |
| Gene Accession No. | Q15077 |
| Gene Alias | 2010204J23Rik; G-coupled nucleotide receptor; MGC15335; P2 purinoceptor; P2ry6; P2Y ATP receptor 6; P2Y purinoceptor 6; p2y3 and chick; p2y3 receptor; P2Y6; P2Y6 receptor; PP2891; Purinoceptor p2y6; pyrimidinergic receptor P2Y, G-protein coupled, 6; pyrimidinergic receptor P2Y6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.