Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2Y8 Monoclonal Antibody (1G5A11)
GREENER_CHOICE

Catalog No. PIMA531824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531824 Supplier Invitrogen™ Supplier No. MA531824
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Amino acid sequence of the immunogen is proprietary. This antibody has been tested in indirect ELISA.

P2Y8 belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. The gene codifying for this protein is moderately expressed in undifferentiated HL60 cells, and is located at both chromosomes X and Y.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y8
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1G5A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene P2RY8
Gene Accession No. Q86VZ1
Gene Alias G-protein coupled purinergic receptor P2Y8; MGC50878; P2RY8; P2Y purinoceptor 8; P2Y8; purinergic receptor P2Y, G-protein coupled, 8; purinergic receptor P2Y8; RP11-261P4.4
Gene Symbols P2RY8
Host Species Mouse
Immunogen Purified recombinant fragment of human P2RY8 expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286530
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.