Learn More
Velp™ P3 Glass Crucibles for FIWE3 Fiber Analyzer
SDP
Catalog No. 11392027
Change view
Click to view available options
Porosity:
Class 3
Catalog No. Porosity
11-392-027 Class 3
1 options
This item is not returnable. View return policy
Catalog No. 11-392-027 Supplier Velp™ Supplier No. A00000137
Add to Cart
Add to Cart
This item is not returnable. View return policy
P3 glass crucibles for use with the FIWE3 Fiber Analyzer.
  • Glass crucibles, P3
  • For fiber analyzer
Specifications
Material Glass
Quantity 6 Crucibles
Capacity (Metric) 16 to 40 microns
Autoclavable Not Autoclavable
Product Type Glass Crucible
Porosity Class 3
Shape Round