Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ P30

Catalog No. 89955500 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-955-500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-955-500 Supplier Abnova™ Supplier No. P5177
Add to Cart
Add to Cart

Used for SDS-PAGE

Specifications

Accession Number Q27298
For Use With (Application) SDS-PAGE
Formulation Lyophilized
Molecular Weight (g/mol) 43kDa
Name P30 (Toxoplasma Gondii) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 500 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with distilled water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name P30
Species E. coli
Protein Tag GST
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.