Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p35 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990888
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0888 100 μg
50-199-0889 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0888 Supplier Rockland Immunochemicals Supplier No. 200401C12
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for Borrelia burgdorferi p35 protein. A BLAST analysis was used to suggest cross-reactivity with p35 from B. burgdorferi, garinii, and afzelii sources based on 100% homology with the immunizing sequence. Cross-reactivity with p35 from other sources has not been determined.

The protein encoded by this gene (p35) is a neuron-specific activator of cyclin-dependent kinase 5 (CDK5); the activation of CDK5 is required for proper development of the central nervous system. The p35 form of this protein is proteolytically cleaved by calpain, generating a p25 form. The cleavage of p35 into p25 results in relocalization of the protein from the cell periphery to nuclear and perinuclear regions. P25 deregulates CDK5 activity by prolonging its activation and changing its cellular location. The p25 form accumulates in the brain neurons of patients with Alzheimer's disease. This accumulation correlates with an increase in CDK5 kinase activity, and may lead to aberrantly phosphorylated forms of the microtubule-associated protein tau, which contributes to Alzheimer's disease.

Specifications

Antigen p35
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Alias CDK5 activator 1; CDK5P35; Cdk5r; Cdk5r1; cyclin dependent kinase 5 regulatory subunit 1; Cyclin-dependent kinase 5 activator 1; Cyclin-dependent kinase 5 activator 1, p25; Cyclin-dependent kinase 5 activator 1, p35; Cyclin-dependent kinase 5 regulatory subunit 1; cyclin-dependent kinase 5, regulatory subunit (p35) 1; cyclin-dependent kinase 5, regulatory subunit 1 (p35); D11Bwg0379e; Nck5a; neuronal CDK5 activator; p23; p25; p35; p35nck5a; regulatory partner for CDK5 kinase; Tau protein kinase II 23 kDa subunit; tau protein kinase II 23kDa subunit; TPKII regulatory subunit
Host Species Rabbit
Immunogen MBP-fusion protein corresponding to Borrelia burgdorferi p35 protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.