Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p38 MAPK Monoclonal Antibody (p38-3F11)
GREENER_CHOICE

Catalog No. 331300
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-130-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-130-0 Supplier Invitrogen™ Supplier No. 331300
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

33-1300 has been successfully used in immunofluorescence, and western blot with human recombinant proteins and whole cell lysates of hamster, mouse, and primate origins. 33-1300 specifically recognizes both alpha and beta isoforms of p38, with slight reactivity to p38-delta.

The p38 MAPK cascade regulates a variety of cellular responses to stress, inflammation, and other signals. p38 MAPK is relatively inactive in the nonphosphorylated form and becomes rapidly activated by dual phosphorylation of a Thr-Gly-Tyr motifs. There are four isoforms of p38 MAPK, which differ in their tissue expression and affinity for upstream activators and downstream effectors. When cells are exposed to tumor necrosis factor, interleukin-1, heat shock, or other activating stimuli, activation of MAPK kinase-3 occurs by phosphorylation. Activated MAPK kinase-3/6 phosphorylate each residue of Thr180 and Tyr182 in p38 MAPK. Phospho-p38 MAPK activates ATF-2, CHOP-1, MEF-2 and other transcription factors through phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p38 MAPK
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone p38-3F11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene MAPK14
Gene Accession No. P47811, P70618, Q16539
Gene Alias Crk1; CSAID-binding protein; Csaids binding protein; CSBP; CSBP1; CSBP2; CSPB1; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; Exip; Hog; MAP kinase 11; MAP kinase 13; MAP kinase 14; MAP kinase 2; MAP kinase Mxi2; MAP kinase p38 alpha; MAP kinase p38 beta; MAP kinase p38 delta; MAPK 11; MAPK 13; MAPK 14; Mapk11; Mapk13; MAPK-13; Mapk14; MAX-interacting protein 2; mitogen activated protein kinase 11; mitogen activated protein kinase 13; mitogen activated protein kinase 14; mitogen-activated protein kinase 11; Mitogen-activated protein kinase 13; mitogen-activated protein kinase 14; mitogen-activated protein kinase 14A; mitogen-activated protein kinase p38 alpha; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38 delta; mitogen-activated protein kinase p38-2; MXI2; p38; p38 alpha; p38 delta MAP kinase; p38 MAP kinase; p38 MAP kinase alpha; p38 MAPK; p38 mitogen activated protein kinase; p38-2; p38a; p38ALPHA; p38-alpha; p38alpha Exip; P38b; p38Beta; p38beta2; p38delta; p38Hog; p38MAPK; Prkm11; PRKM13; PRKM14; Prkm15; protein kinase, mitogen activated kinase, 11, p38beta; reactive kinase; RK; SAPK/Erk/kinase 4; SAPK2; SAPK2A; SAPK2B; SAPK4; Serk4; Stress-activated protein kinase 2a; Stress-activated protein kinase 2b; Stress-activated protein kinase 4; stress-activated protein kinase-2; stress-activated protein kinase-2b; tRNA synthetase cofactor p38
Gene Symbols MAPK14
Host Species Mouse
Immunogen Full-length recombinant mouse p38 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1432, 26416, 81649
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
What is the function of p38MAPK?

p38 mitogen-activated protein kinase (MAPK) phosphorylates target proteins at serine and threonine residues in response to stress stimuli such as UV, heat shock, and osmotic shock which often leads to apoptosis. Activated p38MAPK can phosphorylate transcription factors including ATF2, Mac, and MEF2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.