Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ p38 MAPK InhibitorTocris Bioscience™et™

Catalog No. 22441R
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-441-R 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-441-R Supplier Tocris Bioscience™ Supplier No. 2244/1 SET
This item is not returnable. View return policy

Selection of 3 p38 MAPK inhibitors (Cat. Nos. 1264, 1202 and 1962)

Contains the listed products which can be used to selectively inhibit p38 MAPK signalling. The products are pre-dissolved in DMSO at a stock concentration of 10 mM in 0.5 ml.

Specifications

Purity >95%
Content And Storage -20°C
Quantity 1 Set
Product Type p38 MAPK Inhibitor Tocriset™
Inhibitors p38 MAPK Inhibitor Tocriset™

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.