Learn More
Biocare Medical, LLC p40 (M) Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC API3066AA
Encompass_Preferred
The mouse monoclonal antibody p40 [BC28] recognizes an epitope unique to the p40 protein and may have applications in cases where p63 has traditionally been used. p63 [4A4] recognizes both the p63 and p40 proteins. As a result, p63 suffers from specificity limitations due to reactivity in a subset of lung adenocarcinomas (ADC). In contrast, p40 is selectively expressed in lung Squamous cell carcinoma (SqCC), offering an opportunity for improved specificity. p40 antibody (M) [BC28] recognizes an epitope unique to p40, which may result in diminished reactivity in lung ADC and increased specificity. Studies have supported routine use of p40 as an alternative for p63. In contrast to the rabbit polyclonal p40, p40 [BC28] does not stain macrophages.
Catalog No. NC0457680
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.