Learn More
Diagnostic Biosystems P504S/ALPHA-METHYLACYL-COA p504S/Alpha-Methylacyl-CoA Racemase (AMACR)
Supplier:  Diagnostic Biosystems RMAB07805
Encompass_Preferred
P504S (AMACR) is an essential enzyme in the β-oxidation of branched-chain fatty acids. Recently, P504S was identified through cDNA library subtraction and microarrays in malignant prostate tissues. High expression of P504S protein is found in prostatic adenocarcinoma but not in benign prostatic tissue by immunohistochemical staining in paraffin-embedded tissues. The expression of P504S is also detected in two premalignant lesions of the prostate: high-grade prostatic intraepithelial neoplasia (PIN) and atypical adenomatous hyperplasia. Using P504S as a positive marker along with basal cell staining (34 βE12 or P63) as a negative marker could help to confirm the diagnosis of small focus of prostate carcinoma on needle biopsy.
Catalog No. 50-238-5311
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.