Learn More
Biocare Medical, LLC P504S Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC ACA200C
Encompass_Preferred
P504S, also known as α-methylacyl coenzyme A racemase (AMACR), is a peroxisomal and mitochondrial enzyme that plays a role in bile acid synthesis and β-oxidation of branched chain fatty acids. P504S was initially identified from a cDNA library as a gene that is overexpressed in human prostate cancer; with little or no expression in normal prostate. In immunohistochemistry, P504S (P) antibody has been shown to be a specific marker of prostatic adenocarcinoma. Additionally, prostate glands involved in PIN have been found to express P504S, whereas P504S was nearly undetectable in benign glands.
Catalog No. 50-827-73
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.