Learn More
TARGETMOL CHEMICALS INC OXYFEDRINE L-FORM HCL 5MG
Supplier: TARGETMOL CHEMICALS INC T24579
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Oxyfedrine L-form HCl (Ildamen) is a partial agonist at beta-adrenergic receptors and acts as a coronary vasodilator and cardiotonic agent. purity: 98%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.